accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1267603-73-5

IUPAC Name: 2-{4-[2-(5-{2-[2-(benzyloxy)-4-(dibutylamino)phenyl]ethenyl}thiophen-2-yl)ethenyl]-3-cyano-5-phenyl-5-(trifluoromethyl)-2,5-dihydrofuran-2-ylidene}propanedinitrile
Synonyms: 2-{4-[2-(5-{2-[2-(benzyloxy)-4-(dibutylamino)phenyl]ethenyl}thiophen-2-yl)ethenyl]-3-cyano-5-phenyl-5-(trifluoromethyl)furan-2-ylidene}propanedinitrile
Molecular Weight (g/mol): 744.88
Molecular Formula: C44H39F3N4O2S
InChi Key: SXFGKYGUYHUANQ-UHFFFAOYNA-N
SMILES: CCCCN(CCCC)C1=CC(OCC2=CC=CC=C2)=C(C=CC2=CC=C(S2)C=CC2=C(C#N)C(OC2(C2=CC=CC=C2)C(F)(F)F)=C(C#N)C#N)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results