accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 126807-08-7

IUPAC Name: 3-bromo-4-nitro-1H-indole
Synonyms: 3-bromo-4-nitro-1H-indole
Molecular Weight (g/mol): 241.04
Molecular Formula: C8H5BrN2O2
InChi Key: VYQRPDVMVMLVHA-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C2C(Br)=CNC2=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)