accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOOOOOONOOOONaNaNaNa

CAS RN 126824-24-6

IUPAC Name: tetrasodium 2-{[2-(2-{2-[bis(carboxylatomethyl)amino]phenoxy}ethoxy)phenyl](carboxylatomethyl)amino}acetate
Synonyms: tetrasodium 2-{[2-(2-{2-[bis(carboxylatomethyl)amino]phenoxy}ethoxy)phenyl](carboxylatomethyl)amino}acetate
Molecular Weight (g/mol): 564.37
Molecular Formula: C22H20N2Na4O10
InChi Key: ZWSMLJACYSGFKD-UHFFFAOYSA-J
SMILES: [Na+].[Na+].[Na+].[Na+].[O-]C(=O)CN(CC([O-])=O)C1=CC=CC=C1OCCOC1=CC=CC=C1N(CC([O-])=O)CC([O-])=O
Physical Form 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Grade 
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results