accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOOHHNOOOCH3

CAS RN 126828-35-1

IUPAC Name: 2-{4-[(2,4-dimethoxyphenyl)({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)methyl]phenoxy}acetic acid
Synonyms: 4-[(2,4-dimethoxyphenyl)({[(9H-fluoren-9-ylmethoxy)carbonyl]amino})methyl]phenoxyacetic acid
Molecular Weight (g/mol): 539.58
Molecular Formula: C32H29NO7
InChi Key: UPMGJEMWPQOACJ-UHFFFAOYNA-N
SMILES: COC1=CC(OC)=C(C=C1)C(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C1=CC=C(OCC(O)=O)C=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)