accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 126833-17-8

IUPAC Name: N-(2,3-dichloro-4-hydroxyphenyl)-1-methylcyclohexane-1-carboxamide
Synonyms: fenhexamid
Molecular Weight (g/mol): 302.2
Molecular Formula: C14H17Cl2NO2
InChi Key: VDLGAVXLJYLFDH-UHFFFAOYSA-N
SMILES: CC1(CCCCC1)C(=O)NC1=CC=C(O)C(Cl)=C1Cl
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results