accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1268519-74-9

IUPAC Name: 9,9-diphenyl-9H-fluoren-2-amine
Synonyms: 9,9-diphenylfluoren-2-amine
Molecular Weight (g/mol): 333.43
Molecular Formula: C25H19N
InChi Key: MQRGCMXCVJPWHI-UHFFFAOYSA-N
SMILES: NC1=CC2=C(C=C1)C1=CC=CC=C1C2(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results