accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFSOOOFFFSOOOFFFSOOOLu

CAS RN 126857-69-0

IUPAC Name: lutetium(3+) tritrifluoromethanesulfonate
Synonyms: lutetium(3+) tritriflate
Molecular Weight (g/mol): 622.16
Molecular Formula: C3F9LuO9S3
InChi Key: NRMNRSCGHRWJAK-UHFFFAOYSA-K
SMILES: [Lu+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results