accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3COHNNNHONOHNOOHNHOHONHOCH3H3CHNONNH2H2NH2N

CAS RN 12687-51-3

IUPAC Name: 2-{[1-(2-{2-[2-(2-{2-amino-5-[(diaminomethylidene)amino]pentanamido}-3-methylbutanamido)-3-(4-hydroxyphenyl)propanamido]-3-methylpentanamido}-3-(1H-imidazol-5-yl)propanoyl)pyrrolidin-2-yl]formamido}-3-phenylpropanoic acid
Synonyms: 2-{[1-(2-{2-[2-(2-{2-amino-5-[(diaminomethylidene)amino]pentanamido}-3-methylbutanamido)-3-(4-hydroxyphenyl)propanamido]-3-methylpentanamido}-3-(3H-imidazol-4-yl)propanoyl)pyrrolidin-2-yl]formamido}-3-phenylpropanoic acid
Molecular Weight (g/mol): 931.11
Molecular Formula: C46H66N12O9
InChi Key: QMMRCKSBBNJCMR-UHFFFAOYNA-N
SMILES: CCC(C)C(NC(=O)C(CC1=CC=C(O)C=C1)NC(=O)C(NC(=O)C(N)CCCN=C(N)N)C(C)C)C(=O)NC(CC1=CN=CN1)C(=O)N1CCCC1C(=O)NC(CC1=CC=CC=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)