CAS RN 1269669-43-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
exo-DNTT-PMI (DNTT-Precursor) 95.0+%, TCI America™
CAS: 1269669-43-3 Molecular Formula: C32H19NO2S2 Synonym: exo-Dinaphthothienothiophene-phenylmaleimide Monoadduct, exo-Naphtho[2,3-b]naphtho[2′,3′:4,5]thieno[2,3-d]thiophene N-Phenylmaleimide Monoadduct