accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 127-20-8

IUPAC Name: sodium 2,2-dichloropropanoate
Synonyms: dalapon (sodium salt)
Molecular Weight (g/mol): 164.94
Molecular Formula: C3H3Cl2NaO2
InChi Key: PDEFQWNXOUGDJR-UHFFFAOYSA-M
SMILES: [Na+].CC(Cl)(Cl)C([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results