accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OH3CCH3CH3(E)

CAS RN 127-41-3

IUPAC Name: (3E)-4-(2,6,6-trimethylcyclohex-2-en-1-yl)but-3-en-2-one
Synonyms: ionone α-ionone (natural) cyclocitrylideneacetone, α- α-cyclocitrylideneacetone
Molecular Weight (g/mol): 192.3
Molecular Formula: C13H20O
InChi Key: UZFLPKAIBPNNCA-BQYQJAHWNA-N
SMILES: CC(=O)\C=C\C1C(C)=CCCC1(C)C
Boiling Point 
  • (2)
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)