accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaCH3ONSOONH2

CAS RN 127-56-0

IUPAC Name: sodium acetyl(4-aminobenzenesulfonyl)azanide
Synonyms: sulfacetamide sodium sodium sulfanilylacetamide sulfacetamide sodium anhydrous sodium N-sulfanilylacetamide
Molecular Weight (g/mol): 236.22
Molecular Formula: C8H9N2NaO3S
InChi Key: PQMSFAORUFMASU-UHFFFAOYSA-M
SMILES: [Na+].CC(=O)[N-]S(=O)(=O)C1=CC=C(N)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results