accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNHSOOH2NN

CAS RN 127-79-7

IUPAC Name: 4-amino-N-(4-methylpyrimidin-2-yl)benzene-1-sulfonamide
Synonyms: sulfamerazine veyx brand of sulfamerazine pirimal-M mesulfa sulfameradine romezin 2-sulfa-4-methylpyrimidine cremomerazine sulfamerazine preparation methylsulfazin
Molecular Weight (g/mol): 264.3
Molecular Formula: C11H12N4O2S
InChi Key: QPPBRPIAZZHUNT-UHFFFAOYSA-N
SMILES: CC1=CC=NC(NS(=O)(=O)C2=CC=C(N)C=C2)=N1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results