accessibility menu, dialog, popup

UserName

Viewing profile as user:
NiHCHC

CAS RN 1271-28-9

IUPAC Name: nickel(2+) bis(cyclopenta-2,4-dien-1-ide)
Synonyms: nickelocene
Molecular Weight (g/mol): 188.88
Molecular Formula: C10H10Ni
InChi Key: KZPXREABEBSAQM-UHFFFAOYSA-N
SMILES: [Ni++].[CH-]1C=CC=C1.[CH-]1C=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (5)
Quantity 
  • (1)
  • (1)
  • (3)
  • (1)
Physical Form 
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (1)
  • (5)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)