accessibility menu, dialog, popup

UserName

Viewing profile as user:
FeOOH

CAS RN 1271-42-7

IUPAC Name: Ferrocenecarboxylic acid
Synonyms: —
Molecular Weight (g/mol): 230.04
Molecular Formula: C11H10FeO2
InChi Key: —
SMILES: [Fe].c1cccc1.OC(=O)c1cccc1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)