accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH4CH4Ti

CAS RN 1271-66-5

IUPAC Name: bis(cyclopenta-1,3-diene) bis(methane) titanium
Synonyms: bis(cyclopentadiene) bis(methane) titanium
Molecular Weight (g/mol): 212.16
Molecular Formula: C12H20Ti
InChi Key: CRKLPNOLYBORCP-UHFFFAOYSA-N
SMILES: C.C.[Ti].C1C=CC=C1.C1C=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results