accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1272-22-6

IUPAC Name: holmium(3+) tris(cyclopenta-2,4-dien-1-ide)
Synonyms: holmium(3+) tris(cyclopentadienide)
Molecular Weight (g/mol): 360.22
Molecular Formula: C15H15Ho
InChi Key: FQEKFYCUZKBSQY-UHFFFAOYSA-N
SMILES: [Ho+3].[CH-]1C=CC=C1.[CH-]1C=CC=C1.[CH-]1C=CC=C1
Boiling Point 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)