accessibility menu, dialog, popup

UserName

Viewing profile as user:
OBr

CAS RN 127264-14-6

IUPAC Name: 5-(2-bromoethyl)-2,3-dihydro-1-benzofuran
Synonyms: 5-(2-bromoethyl)-2,3-dihydro-1-benzofuran
Molecular Weight (g/mol): 227.1
Molecular Formula: C10H11BrO
InChi Key: JRKZQRRYNCMSCB-UHFFFAOYSA-N
SMILES: BrCCC1=CC2=C(OCC2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results