accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3NNH2

CAS RN 127285-08-9

IUPAC Name: 1-(propan-2-yl)piperidin-4-amine
Synonyms: 1-isopropylpiperidin-4-amine
Molecular Weight (g/mol): 142.25
Molecular Formula: C8H18N2
InChi Key: ZRQQXFMGYSOKDF-UHFFFAOYSA-N
SMILES: CC(C)N1CCC(N)CC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results