CAS RN 127294-77-3
CAS RN 127294-77-3
IUPAC Name:
N-methylpiperidin-3-amine dihydrochloride
Synonyms:
N-methylpiperidin-3-amine dihydrochloride
Molecular Weight (g/mol):
187.11
Molecular Formula:
C6H16Cl2N2
InChi Key:
VEXIRMPBAHTVNL-UHFFFAOYNA-N
SMILES:
Cl.Cl.CNC1CCCNC1
1
–
1
of
1
results
1
3-(Methylamino)piperidine Dihydrochloride 98.0+%, TCI America™
CAS: 127294-77-3 Molecular Formula: C6H16Cl2N2 Molecular Weight (g/mol): 187.108 MDL Number: MFCD00673143 InChI Key: VEXIRMPBAHTVNL-UHFFFAOYSA-N PubChem CID: 24820174 IUPAC Name: N-methylpiperidin-3-amine;dihydrochloride SMILES: CNC1CCCNC1.Cl.Cl