accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1273-89-8

IUPAC Name: λ²-iron(2+) 1-ethylcyclopenta-2,4-dien-1-ide cyclopenta-2,4-dien-1-ide
Synonyms: ethylferrocene
Molecular Weight (g/mol): 214.09
Molecular Formula: C12H14Fe
InChi Key: FCNXGBYXGSKCDG-UHFFFAOYSA-N
SMILES: [Fe++].[CH-]1C=CC=C1.CC[C-]1C=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results