accessibility menu, dialog, popup

UserName

Viewing profile as user:
FeHCHCCHCHCCH3OCCH3O

CAS RN 1273-94-5

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 270.11
Molecular Formula: C14H14FeO2
InChi Key: DAVCBKPWWUUYNL-UHFFFAOYSA-N
SMILES: [Fe].CC(=O)[C-]1[CH-][CH-][CH-][CH-]1.CC(=O)[c-]1cccc1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results