accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 127408-31-5

IUPAC Name: N-[(3S,4R)-6-cyano-3-hydroxy-2,2-dimethyl-3,4-dihydro-2H-1-benzopyran-4-yl]-N-hydroxyacetamide
Synonyms: N-[(3S,4R)-6-cyano-3-hydroxy-2,2-dimethyl-3,4-dihydro-1-benzopyran-4-yl]-N-hydroxyacetamide
Molecular Weight (g/mol): 276.29
Molecular Formula: C14H16N2O4
InChi Key: IWTCFIIOUXJOOV-OLZOCXBDSA-N
SMILES: CC(=O)N(O)[C@H]1[C@H](O)C(C)(C)OC2=CC=C(C=C12)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results