accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 12741-05-8

IUPAC Name: yttrium(3+) trichloride
Synonyms: yttrium chloride|yttrium(III) chloride|yttrium trichloride|YCl3
Molecular Weight (g/mol): 195.26
Molecular Formula: Cl3Y
InChi Key: PCMOZDDGXKIOLL-UHFFFAOYSA-K
SMILES: [Cl-].[Cl-].[Cl-].[Y+3]
Grade 
  • (7)
Percent Purity 
  • (3)
  • (4)
Physical Form 
  • (4)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (7)
  • (3)
  • (4)
  • (4)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)