accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOONHOO

CAS RN 127582-76-7

IUPAC Name: 7-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)heptanoic acid
Synonyms: 7-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}heptanoic acid
Molecular Weight (g/mol): 367.45
Molecular Formula: C22H25NO4
InChi Key: FRPYXTFBPCYALT-UHFFFAOYSA-N
SMILES: OC(=O)CCCCCCNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results