accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NCH3NCH3H3CSOOOSOOONa

CAS RN 12768-78-4

IUPAC Name: sodium 4-{[4-(diethylamino)phenyl][4-(diethyliminiumyl)cyclohexa-2,5-dien-1-ylidene]methyl}naphthalene-2,7-disulfonate
Synonyms: sodium 4-{[4-(diethylamino)phenyl][4-(diethyliminio)cyclohexa-2,5-dien-1-ylidene]methyl}naphthalene-2,7-disulfonate
Molecular Weight (g/mol): 616.72
Molecular Formula: C31H33N2NaO6S2
InChi Key: UWGCNDBLFSEBDW-UHFFFAOYSA-M
SMILES: [Na+].CCN(CC)C1=CC=C(C=C1)C(=C1C=CC(C=C1)=[N+](CC)CC)C1=CC(=CC2=CC(=CC=C12)S([O-])(=O)=O)S([O-])(=O)=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results