CAS RN 127783-34-0
CAS RN 127783-34-0
IUPAC Name:
ethynyl(phenyl)iodanium; tetrafluoroboranuide
Synonyms:
ethynyl(phenyl)iodanium tetrafluoroborate
Molecular Weight (g/mol):
315.84
Molecular Formula:
C8H6BF4I
InChi Key:
AVSBPVBKCONUGF-UHFFFAOYSA-N
SMILES:
F[B-](F)(F)F.C#C[I+]C1=CC=CC=C1
1
–
1
of
1
results
1
Ethynyl(phenyl)iodonium Tetrafluoroborate 97.0+%, TCI America™
CAS: 127783-34-0 Molecular Formula: C8H6BF4I Molecular Weight (g/mol): 315.844 MDL Number: MFCD00191648 InChI Key: AVSBPVBKCONUGF-UHFFFAOYSA-N PubChem CID: 2758943 IUPAC Name: ethynyl(phenyl)iodanium;tetrafluoroborate SMILES: [B-](F)(F)(F)F.C#C[I+]C1=CC=CC=C1