accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOHHNBrCl

CAS RN 127792-23-8

IUPAC Name: 2-{2-[(2-bromo-6-chlorophenyl)amino]phenyl}acetic acid
Synonyms: {2-[(2-bromo-6-chlorophenyl)amino]phenyl}acetic acid
Molecular Weight (g/mol): 340.6
Molecular Formula: C14H11BrClNO2
InChi Key: RABKUCPRNJPBRH-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CC=CC=C1NC1=C(Cl)C=CC=C1Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results