accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 127988-21-0

IUPAC Name: 2-[(1H-1,2,4-triazol-1-yl)methyl]aniline
Synonyms: 2-(1,2,4-triazol-1-ylmethyl)aniline
Molecular Weight (g/mol): 174.21
Molecular Formula: C9H10N4
InChi Key: CBGQHZNXHUCAGE-UHFFFAOYSA-N
SMILES: NC1=CC=CC=C1CN1C=NC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results