accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 128-62-1

IUPAC Name: (3S)-6,7-dimethoxy-3-[(5R)-4-methoxy-6-methyl-2H,5H,6H,7H,8H-[1,3]dioxolo[4,5-g]isoquinolin-5-yl]-1,3-dihydro-2-benzofuran-1-one
Synonyms: noscapine noskapin coscopin tuscalman noscapina noscapect noscapinum α-narcotine longactin key-tusscapine
Molecular Weight (g/mol): 413.43
Molecular Formula: C22H23NO7
InChi Key: AKNNEGZIBPJZJG-MSOLQXFVSA-N
SMILES: COC1=CC=C2[C@H](OC(=O)C2=C1OC)[C@@H]1N(C)CCC2=CC3=C(OCO3)C(OC)=C12
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)