accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CFFOOH

CAS RN 128073-33-6

IUPAC Name: 2,2-difluoro-1-methylcyclopropane-1-carboxylic acid
Synonyms: 2,2-difluoro-1-methylcyclopropane-1-carboxylic acid
Molecular Weight (g/mol): 136.1
Molecular Formula: C5H6F2O2
InChi Key: HLFLYOQLHYYNLT-UHFFFAOYNA-N
SMILES: CC1(CC1(F)F)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)