accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3NOON

CAS RN 128119-95-9

IUPAC Name: 8-(diethylamino)-12-oxo-12H-10-oxa-5-azatetraphene-11-carbonitrile
Synonyms: 8-(diethylamino)-12-oxo-10-oxa-5-azatetraphene-11-carbonitrile
Molecular Weight (g/mol): 343.39
Molecular Formula: C21H17N3O2
InChi Key: DOIVGWIGBQTTHP-UHFFFAOYSA-N
SMILES: CCN(CC)C1=CC=C2N=C3C(OC2=C1)=C(C#N)C(=O)C1=CC=CC=C31
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results