CAS RN 128143-89-5
4'-Chloro-2,2':6',2″-terpyridine, 99%
CAS: 128143-89-5 Molecular Formula: C15H10ClN3 Molecular Weight (g/mol): 267.72 MDL Number: MFCD00191930 InChI Key: AHEMFMCEBIJRMU-UHFFFAOYSA-N Synonym: 4'-chloro-2,2':6',2-terpyridine,4'-chloro-2,2':6',2'-terpyridine,4'-chloro-2,2',6',2-terpyridine,4-chloro-2,6-bis pyridin-2-yl pyridine,4-chloro-2,6-di 2-pyridyl pyridine,4-chloro-6-pyridin-2-yl-2,2'-bipyridine,zlchem 975,acmc-1cg8d,4'-chloro-2,2'6'2-terpyridine,4-chloro-2,6-bis 2-pyridyl pyridine PubChem CID: 667748 IUPAC Name: 4-chloro-2,6-dipyridin-2-ylpyridine SMILES: ClC1=CC(=NC(=C1)C1=CC=CC=N1)C1=CC=CC=N1
4'-Chloro-2,2':6',2″-terpyridine, 98%
CAS: 128143-89-5 Molecular Formula: C15H10ClN3 Molecular Weight (g/mol): 267.72 MDL Number: MFCD00191930 InChI Key: AHEMFMCEBIJRMU-UHFFFAOYSA-N Synonym: 4'-chloro-2,2':6',2-terpyridine,4'-chloro-2,2':6',2'-terpyridine,4'-chloro-2,2',6',2-terpyridine,4-chloro-2,6-bis pyridin-2-yl pyridine,4-chloro-2,6-di 2-pyridyl pyridine,4-chloro-6-pyridin-2-yl-2,2'-bipyridine,zlchem 975,acmc-1cg8d,4'-chloro-2,2'6'2-terpyridine,4-chloro-2,6-bis 2-pyridyl pyridine PubChem CID: 667748 SMILES: ClC1=CC(=NC(=C1)C1=CC=CC=N1)C1=CC=CC=N1
4'-Chloro-2,2':6',2″-terpyridine 98.0+%, TCI America™
CAS: 128143-89-5 Molecular Formula: C15H10ClN3 Molecular Weight (g/mol): 267.72 MDL Number: MFCD00191930 InChI Key: AHEMFMCEBIJRMU-UHFFFAOYSA-N Synonym: 4'-chloro-2,2':6',2-terpyridine,4'-chloro-2,2':6',2'-terpyridine,4'-chloro-2,2',6',2-terpyridine,4-chloro-2,6-bis pyridin-2-yl pyridine,4-chloro-2,6-di 2-pyridyl pyridine,4-chloro-6-pyridin-2-yl-2,2'-bipyridine,zlchem 975,acmc-1cg8d,4'-chloro-2,2'6'2-terpyridine,4-chloro-2,6-bis 2-pyridyl pyridine PubChem CID: 667748 IUPAC Name: 4-chloro-6-(pyridin-2-yl)-2,2'-bipyridine SMILES: ClC1=CC(=NC(=C1)C1=CC=CC=N1)C1=CC=CC=N1
Sigma Aldrich 4'-Chloro-2,2':6',2''-terpyridine
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.