accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 128201-89-8

IUPAC Name: 4-(icosa-5,8,11,14-tetraenamido)butanoic acid
Synonyms: 4-(icosa-5,8,11,14-tetraenamido)butanoic acid
Molecular Weight (g/mol): 389.58
Molecular Formula: C24H39NO3
InChi Key: JKUDIEXTAYKJNX-UHFFFAOYSA-N
SMILES: CCCCCC=CCC=CCC=CCC=CCCCC(=O)NCCCC(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results