accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COON

CAS RN 128259-47-2

IUPAC Name: ethyl 1-benzyl-1H-pyrrole-3-carboxylate
Synonyms: ethyl 1-benzylpyrrole-3-carboxylate
Molecular Weight (g/mol): 229.28
Molecular Formula: C14H15NO2
InChi Key: KSZAVNAKDOXZGJ-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CN(CC2=CC=CC=C2)C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results