accessibility menu, dialog, popup

UserName

Viewing profile as user:
BHOOH

CAS RN 128388-54-5

IUPAC Name: {5-phenyl-[1,1'-biphenyl]-3-yl}boronic acid
Synonyms: 5-phenyl-[1,1'-biphenyl]-3-ylboronic acid
Molecular Weight (g/mol): 274.13
Molecular Formula: C18H15BO2
InChi Key: MRBZYVMZUBUDAX-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(=CC(=C1)C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results