accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 128453-98-5

IUPAC Name: 4-[(tert-butoxy)carbonyl]thiomorpholine-3-carboxylic acid
Synonyms: 4-(tert-butoxycarbonyl)thiomorpholine-3-carboxylic acid
Molecular Weight (g/mol): 247.31
Molecular Formula: C10H17NO4S
InChi Key: CTDIKDIZNAGMFK-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)N1CCSCC1C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results