accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 128562-95-8

IUPAC Name: 4-(trifluoromethyl)-1H-indole
Synonyms: 4-(trifluoromethyl)-1H-indole
Molecular Weight (g/mol): 185.15
Molecular Formula: C9H6F3N
InChi Key: YTVBZSLUNRYKID-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=C2C=CNC2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)