accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 128733-85-7

IUPAC Name: (5-tert-butyl-2-methoxyphenyl)boronic acid
Synonyms: 5-tert-butyl-2-methoxyphenylboronic acid
Molecular Weight (g/mol): 208.06
Molecular Formula: C11H17BO3
InChi Key: LXFOJKCQSAYVMF-UHFFFAOYSA-N
SMILES: COC1=C(C=C(C=C1)C(C)(C)C)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results