accessibility menu, dialog, popup

UserName

Viewing profile as user:
SOBr

CAS RN 128746-80-5

IUPAC Name: 2-bromo-1-[4-(thiophen-2-yl)phenyl]ethan-1-one
Synonyms: 2-bromo-1-[4-(thiophen-2-yl)phenyl]ethanone
Molecular Weight (g/mol): 281.17
Molecular Formula: C12H9BrOS
InChi Key: QFHKNYPNLSJEQI-UHFFFAOYSA-N
SMILES: BrCC(=O)C1=CC=C(C=C1)C1=CC=CS1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results