CAS RN 128917-74-8
CAS RN 128917-74-8
IUPAC Name:
12-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)dodecanoic acid
Synonyms:
12-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}dodecanoic acid
Molecular Weight (g/mol):
437.58
Molecular Formula:
C27H35NO4
InChi Key:
HVGIKYAQSSNFCH-UHFFFAOYSA-N
SMILES:
OC(=O)CCCCCCCCCCCNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
1
–
1
of
1
results
1
Advanced Chem Tech Fmoc-12-Ado-OH, Advanced ChemTech
CAS: 128917-74-8 Molecular Formula: C27H35NO4 Molecular Weight (g/mol): 437.58 MDL Number: MFCD00235888 InChI Key: HVGIKYAQSSNFCH-UHFFFAOYSA-N IUPAC Name: 12-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)dodecanoic acid SMILES: OC(=O)CCCCCCCCCCCNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12