accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOO

CAS RN 129-64-6

IUPAC Name: 4-oxatricyclo[5.2.1.0²,⁶]dec-8-ene-3,5-dione
Synonyms: carbic anhydride
Molecular Weight (g/mol): 164.16
Molecular Formula: C9H8O3
InChi Key: KNDQHSIWLOJIGP-UHFFFAOYNA-N
SMILES: O=C1OC(=O)C2C3CC(C=C3)C12
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)