accessibility menu, dialog, popup

UserName

Viewing profile as user:
FeOOHOOH

CAS RN 1293-87-4

IUPAC Name: 1,1'-Ferrocenedicarboxylic acid
Synonyms: bis(5-carboxycyclopenta-1,3-dien-1-yl)iron
Molecular Weight (g/mol): 274.05
Molecular Formula: C12H10FeO4
InChi Key: —
SMILES: OC(=O)C1C=CC=C1[Fe]C1=CC=CC1C(O)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)