accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 129332-45-2

IUPAC Name: methyl 3-amino-4-cyano-5-(methylsulfanyl)thiophene-2-carboxylate
Synonyms: methyl 3-amino-4-cyano-5-(methylsulfanyl)thiophene-2-carboxylate
Molecular Weight (g/mol): 228.28
Molecular Formula: C8H8N2O2S2
InChi Key: VBXBCNWDAJLZOC-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(N)C(C#N)=C(SC)S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results