accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1293389-28-2

IUPAC Name: 5,6-difluoro-2,1,3-benzothiadiazole
Synonyms: 5,6-difluoro-2,1,3-benzothiadiazole
Molecular Weight (g/mol): 172.15
Molecular Formula: C6H2F2N2S
InChi Key: ZKBRJEYUJNJNCO-UHFFFAOYSA-N
SMILES: FC1=CC2=NSN=C2C=C1F
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results