accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 129368-70-3

IUPAC Name: [(6-bromohexyl)oxy](tert-butyl)dimethylsilane
Synonyms: [(6-bromohexyl)oxy](tert-butyl)dimethylsilane
Molecular Weight (g/mol): 295.34
Molecular Formula: C12H27BrOSi
InChi Key: PBKXRKYUUXKNSL-UHFFFAOYSA-N
SMILES: CC(C)(C)[Si](C)(C)OCCCCCCBr
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)