accessibility menu, dialog, popup

UserName

Viewing profile as user:
BH4COPNHPRu

CAS RN 1295649-41-0

IUPAC Name: λ²-ruthenium(1+) bis[2-(diphenylphosphanyl)ethyl]amine methanidylidyneoxidanium boranuide
Synonyms: λ²-ruthenium(1+) bis[2-(diphenylphosphanyl)ethyl]amine carbon monoxide boranuide
Molecular Weight (g/mol): 586.42
Molecular Formula: C29H34BNOP2Ru
InChi Key: ZVKXESLAUDGPND-UHFFFAOYSA-N
SMILES: [BH4-].[RuH+].[C-]#[O+].C(CP(C1=CC=CC=C1)C1=CC=CC=C1)NCCP(C1=CC=CC=C1)C1=CC=CC=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results