accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 129604-27-9

IUPAC Name: 1-[3-chloro-4-(trifluoromethoxy)phenyl]ethan-1-one
Synonyms: 1-[3-chloro-4-(trifluoromethoxy)phenyl]ethanone
Molecular Weight (g/mol): 238.59
Molecular Formula: C9H6ClF3O2
InChi Key: BUSQQCHYIYPNTO-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC(Cl)=C(OC(F)(F)F)C=C1
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results