accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNNNOHN

CAS RN 129618-40-2

IUPAC Name: 2-cyclopropyl-7-methyl-2,4,9,15-tetraazatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3(8),4,6,11,13-hexaen-10-one
Synonyms: nevirapine NEV NVP viramune
Molecular Weight (g/mol): 266.3
Molecular Formula: C15H14N4O
InChi Key: NQDJXKOVJZTUJA-UHFFFAOYSA-N
SMILES: CC1=CC=NC2=C1NC(=O)C1=CC=CN=C1N2C1CC1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Grade 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)