accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ClOH

CAS RN 129716-11-6

IUPAC Name: (4-chloro-2-methylphenyl)methanol
Synonyms: (4-chloro-2-methylphenyl)methanol
Molecular Weight (g/mol): 156.61
Molecular Formula: C8H9ClO
InChi Key: AHXBDGJNZJKLNE-UHFFFAOYSA-N
SMILES: CC1=C(CO)C=CC(Cl)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results